Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RalB Antibody (OTI2C4), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245927
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry, Immunofluorescence
RalB Monoclonal specifically detects RalB in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence.Specifications
RalB | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:200 - 1:4000, Immunohistochemistry 1:150, Immunofluorescence 1:100 | |
GTP binding protein, RAS-like protein B, ras-related protein Ral-B, v-ral simian leukemia viral oncogene homolog B (ras related; GTP bindingprotein) | |
Mouse | |
23.2 kDa | |
0.1 mL | |
Signal Transduction | |
5899 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunofluorescence | |
OTI2C4 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
RALB | |
Full length human recombinant protein of human RALB(NP_002872) produced in HEK293T cell. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction