Learn More
Invitrogen™ RALB Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji, PC-12, Rat2, Raw264.7, C2C12. Predicted reactivity: Mouse (95%), Rat (95%), Xenopus laevis (94%), Rhesus Monkey (99%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | RALB |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.71 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | RALB |
| Gene Accession No. | P11234, P36860, Q9JIW9 |
| Gene Alias | 5730472O18Rik; dRalb; GTP binding protein; GTP binding protein); OTTHUMP00000204385; OTTHUMP00000207264; QtsA-18830; Ralb; Ral-b; RALB Ras like proto-oncogene B; RAS like proto-oncogene B; ras related GTP binding protein B; RAS-like protein B; Ras-related protein Ral-B; unnamed protein product; v-ral simian leukemia viral oncogene homolog B; v-ral simian leukemia viral oncogene homolog B (ras related); v-ral simian leukemia viral oncogene homolog B (ras related; GTP binding protein) |
| Gene Symbols | RALB |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.