Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RALB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544575
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544575 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544575 Supplier Invitrogen™ Supplier No. PA544575
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: FREQILRVKA EEDKIPLLVV GNKSDLEERR QVPVEEARSK AEEWGVQYVE Sequence homology: Cow: 100%; Dog: 100%; Goat: 86%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 79%; Zebrafish: 100%.

This gene encodes a GTP-binding protein that belongs to the small GTPase superfamily and Ras family of proteins. GTP-binding proteins mediate the transmembrane signaling initiated by the occupancy of certain cell surface receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RALB
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene RALB
Gene Accession No. P11234
Gene Alias 5730472O18Rik; dRalb; GTP binding protein; GTP binding protein); OTTHUMP00000204385; OTTHUMP00000207264; QtsA-18830; Ralb; Ral-b; RALB Ras like proto-oncogene B; RAS like proto-oncogene B; ras related GTP binding protein B; RAS-like protein B; Ras-related protein Ral-B; unnamed protein product; v-ral simian leukemia viral oncogene homolog B; v-ral simian leukemia viral oncogene homolog B (ras related); v-ral simian leukemia viral oncogene homolog B (ras related; GTP binding protein)
Gene Symbols RALB
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human RALB.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5899
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.