Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    RALBP1 Antibody (OTI11B2), Allophycocyanin, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273788APC
Description
RALBP1 Monoclonal specifically detects RALBP1 in Human, Mouse, Rat, Canine samples. It is validated for Flow Cytometry.Specifications
| RALBP1 | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| RALBP1 | |
| Full length recombinant protein of human RALBP1 (NP_006779) produced in HEK293T cells. | |
| 0.1 mL | |
| 10928.0 | |
| Store at 4°C in the dark. | 
| Flow Cytometry | |
| OTI11B2 | |
| Flow Cytometry | |
| Dinitrophenyl S-glutathione ATPase, ralA binding protein 1, ralA-binding protein 1,76 kDa Ral-interacting protein, RalBP1, Ral-interacting protein 1, RIP, RIP1, RLIP1, RLIP76DNP-SG ATPase | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| IgG1 | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction