Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RALBP1 Antibody (OTI11B2), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273788H
Description
RALBP1 Monoclonal specifically detects RALBP1 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot.Specifications
RALBP1 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
RALBP1 | |
Full length recombinant protein of human RALBP1 (NP_006779) produced in HEK293T cells. | |
0.1 mL | |
10928.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI11B2 | |
Western Blot | |
Dinitrophenyl S-glutathione ATPase, ralA binding protein 1, ralA-binding protein 1,76 kDa Ral-interacting protein, RalBP1, Ral-interacting protein 1, RIP, RIP1, RLIP1, RLIP76DNP-SG ATPase | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Canine | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction