Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RALBP1 Antibody (OTI11B2), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273788MFV500
Description
RALBP1 Monoclonal antibody specifically detects RALBP1 in Human, Mouse, Rat, Canine samples. It is validated for Flow CytometrySpecifications
| RALBP1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| Purified |
| Flow Cytometry | |
| OTI11B2 | |
| Flow Cytometry | |
| Dinitrophenyl S-glutathione ATPase, ralA binding protein 1, ralA-binding protein 1,76 kDa Ral-interacting protein, RalBP1, Ral-interacting protein 1, RIP, RIP1, RLIP1, RLIP76DNP-SG ATPase | |
| Full length human recombinant protein of human RALBP1 (NP_006779) produced in HEK293T cell. | |
| 0.1 mL | |
| Apoptosis, Cell Biology, Immunology, Innate Immunity, Signal Transduction | |
| 10928 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction