Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RALBP1 Monoclonal Antibody (6C6)
Mouse Monoclonal Antibody
$525.50
Specifications
Antigen | RALBP1 |
---|---|
Clone | 6C6 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Immunoprecipitation, Western Blot, Immunocytochemistry |
Description
RALBP1 Monoclonal Antibody for Western Blot, ICC/IF, IP
Can specifically activate hydrolysis of GTP bound to RAC1 and CDC42, but not RALA. Mediates ATP-dependent transport of S-(2,4-dinitrophenyl)-glutathione (DNP-SG) and doxorubicin (DOX) and is the major ATP-dependent transporter of glutathione conjugates of electrophiles (GS-E) and DOX in erythrocytes. Can catalyze transport of glutathione conjugates and xenobiotics, and may contribute to the multidrug resistance phenomenon. Serves as a scaffold protein that brings together proteins forming an endocytotic complex during interphase and also with CDC2 to switch off endocytosis, One of its substrates would be EPN1/Epsin.Specifications
RALBP1 | |
1 mg/mL | |
Immunoprecipitation, Western Blot, Immunocytochemistry | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide | |
Q15311 | |
10928 | |
Full length human recombinant protein of human RALBP1 (NP_006779) produced in HEK293T cell. | |
Antibody |
6C6 | |
-20°C | |
Monoclonal | |
Liquid | |
IgG1 | |
Human, Monkey | |
RALBP1 | |
76 kDa Ral-interacting protein; Cytocentrin; dinitrophenyl S-glutathione ATPase; DNP-SG ATPase; ralA binding protein 1; ralA-binding protein 1; RALBP1; Ral-interacting protein 1; RIP; RIP1; RLIP1; RLIP76 | |
RALBP1 | |
Primary | |
Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title