Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RALBP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543788
Description
Peptide sequence: MTECFLPPTS SPSEHRRVEH GSGLTRTPSS EEISPTKFPG LYRTGEPSPP Sequence homology: Cow: 100%; Dog: 100%; Goat: 86%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
Can specifically activate hydrolysis of GTP bound to RAC1 and CDC42, but not RALA. Mediates ATP-dependent transport of S-(2,4-dinitrophenyl)-glutathione (DNP-SG) and doxorubicin (DOX) and is the major ATP-dependent transporter of glutathione conjugates of electrophiles (GS-E) and DOX in erythrocytes. Can catalyze transport of glutathione conjugates and xenobiotics, and may contribute to the multidrug resistance phenomenon. Serves as a scaffold protein that brings together proteins forming an endocytotic complex during interphase and also with CDC2 to switch off endocytosis, One of its substrates would be EPN1/Epsin.
Specifications
| RALBP1 | |
| Polyclonal | |
| Unconjugated | |
| RALBP1 | |
| 76 kDa Ral-interacting protein; Cytocentrin; dinitrophenyl S-glutathione ATPase; DNP-SG ATPase; ralA binding protein 1; ralA-binding protein 1; RALBP1; Ral-interacting protein 1; RIP; RIP1; RLIP1; RLIP76 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10928 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q15311 | |
| RALBP1 | |
| Synthetic peptide directed towards the N-terminal of human RALBP1 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction