Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RALBP1 Rabbit anti-Human, Unconjugated, Clone: FDI-18, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB20660
Description
RALBP1 Monoclonal antibody specifically detects RALBP1 in Human samples. It is validated for WB-Ce, IHC-P, Flow Cyt.Specifications
RALBP1 | |
Monoclonal | |
Unconjugated | |
RALBP1 | |
Rabbit | |
100 μg | |
10928 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry | |
FDI-18 | |
ralA binding protein 1 | |
RIP1, RLIP1, RLIP76 | |
A synthetic peptide corresponding to human RALBP1 | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction