Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ RalDH2 (ALDH1A2), Rabbit, Unlabeled, Polyclonal,

Catalog No. ABN420MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABN420MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN420MI Supplier MilliporeSigma™ Supplier No. ABN420
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Retinal dehydrogenase 2 (EC 1.2.1.36; UniProt Q62148; also known as Aldehyde dehydrogenase family 1 member A2, RALDH 2, RalDH2, RALDH(II), Retinaldehyde-specific dehydrogenase type 2) is encoded by the Aldh1a2 (also known as Aldh1a7, Raldh2) gene (Gene ID 19378) in murine species. It can recognize and bind both free retinal and cellular retinol-binding protein-bound retinal and oxidize it to retinoate. The enzyme is known to be active in embryos, where retinoic acid play a crucial role in early development. RALDH2-null mice do not live beyond E10.5. The three retinaldehyde dehydrogenases, RALDH1-3 or ALDH1 alpha 1-3, catalyze the irreversible oxidation of retinaldehyde to form atRA. While retinaldehyde is formed via the reversible oxidation of retinol catalyzed by retinol dehydrogenases (RDHs). The three RALDH enzymes cooperatively mediate atRA signaling during eye development through tightly regulated spatiotemporal expression patterns. In the postnatal human eye, RALDH catalytic activity can be detected in the choroid, retinal pigment epithelium (RPE), and retina. RALDH2 is most abundant in the choroid and RPE, moderate abundance in the retina, and in relatively low abundance in sclera. RALDH1 is most abundant in the choroid, in moderate abundance in the sclera, and substantially reduced in the retina and RPE. RALDH3 is most abundantly expressed during embryonic eye development, but not detectable in adult ocular fundal tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RalDH2 (ALDH1A2)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene Aldh1a2, Aldh1a7, Raldh2
Gene Accession No. Q62148
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to an N-terminal region sequence of mouse RalDH2.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_033048
Test Specificity This polyclonal antibody targets an N-terminal region sequence of human/mouse/rat RalDH2 reported by UniProt (O94788/Q62148/Q63639).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.