Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RALGAPA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551467
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551467 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551467 Supplier Invitrogen™ Supplier No. PA551467
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LVSCIQIRSE ENMPGGGLSA GLASANSNVR IIVRDLSGKY SWDSAILYGP PPVSGLSEPT SFMLSLSHQE KPEEPPTSNE CLEDITVKDG LSLQFKRFRE TVPTWDTIRD EEDVLDELLQ YLGVTSPECL QRTGISLNIP APQPVCISE Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 90%.

RALGAPA1 is a catalytic subunit of the heterodimeric RalGAP1 complex which acts as a GTPase activator for the Ras-like small GTPases RALA and RALB.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RALGAPA1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RALGAPA1
Gene Accession No. Q6GYQ0
Gene Alias 2310003F20Rik; 4930400K19Rik; AI563624; GAP-related interacting partner to E12; GAP-related interacting protein to E12; GAP-related-interacting partner to E12; Garnl1; GRIPE; GTPase activating RANGAP domain-like 1; GTPase activating Rap/RanGAP domain-like 1; GTPase-activating Rap/Ran-GAP domain-like 1; GTPase-activating RapGAP domain-like 1; Kiaa0884; mKIAA0884; p240; Ral GTPase activating protein catalytic alpha subunit 1; Ral GTPase activating protein, alpha subunit 1; Ral GTPase activating protein, alpha subunit 1 (catalytic); ral GTPase-activating protein alpha subunit 1; ral GTPase-activating protein subunit alpha-1; Ralgapa1; RalGAPalpha1; Tuberin-like protein 1; tulip 1 protein; TULIP1
Gene Symbols RALGAPA1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RALGAPA1. Recombinant protein control fragment (Product #RP-88983).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 253959
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.