Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RALGAPA1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA568198

Catalog No. PIPA568198


Only null left
Add to Cart

Description

Description

Predicted to react with Mouse and Rat samples.

RALGAPA1 is a catalytic subunit of the heterodimeric RalGAP1 complex which acts as a GTPase activator for the Ras-like small GTPases RALA and RALB.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RALGAPA1
Polyclonal
Unconjugated
RALGAPA1
2310003F20Rik; 4930400K19Rik; AI563624; GAP-related interacting partner to E12; GAP-related interacting protein to E12; GAP-related-interacting partner to E12; Garnl1; GRIPE; GTPase activating RANGAP domain-like 1; GTPase activating Rap/RanGAP domain-like 1; GTPase-activating Rap/Ran-GAP domain-like 1; GTPase-activating RapGAP domain-like 1; Kiaa0884; mKIAA0884; p240; Ral GTPase activating protein catalytic alpha subunit 1; Ral GTPase activating protein, alpha subunit 1; Ral GTPase activating protein, alpha subunit 1 (catalytic); ral GTPase-activating protein alpha subunit 1; ral GTPase-activating protein subunit alpha-1; Ralgapa1; RalGAPalpha1; Tuberin-like protein 1; tulip 1 protein; TULIP1
Rabbit
Affinity Chromatography
RUO
253959, 56784, 56785
-20°C
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O55007, Q6GYP7, Q6GYQ0
RALGAPA1
Synthesized peptide derived from human RALGAPA1.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.