Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RALGAPA1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572292
Description
RALGAPA1 is a catalytic subunit of the heterodimeric RalGAP1 complex which acts as a GTPase activator for the Ras-like small GTPases RALA and RALB.
Specifications
RALGAPA1 | |
Polyclonal | |
Unconjugated | |
RALGAPA1 | |
Ral GTPase-activating protein subunit alpha-1; GAP-related-interacting partner to E12; GRIPE; GTPase-activating Rap/Ran-GAP domain-like 1; Tuberin-like protein 1; p240; RALGAPA1; GARNL1; KIAA0884; TULIP1 | |
Rabbit | |
Protein A | |
RUO | |
253959, 56784 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1.72 mg/mL | |
PBS with 0.09% sodium azide | |
Q6GYP7, Q6GYQ0 | |
RALGAPA1 | |
KLH conjugated synthetic peptide between 735-768 amino acids from human RALGAPA1 | |
200 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction