Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RALGAPA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560441
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560441 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560441 Supplier Invitrogen™ Supplier No. PA560441
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SPAELDLKDD LQQTQGKCRE RQKSESTNSD TTLGCTNEAE LSMGPWQTCE EDPELNTPTD VVADADARHW LQLSPTDA Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 78%.

Catalytic subunit of the heterodimeric RalGAP2 complex which acts as a GTPase activator for the Ras-like small GTPases RALA and RALB.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RALGAPA2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RALGAPA2
Gene Accession No. Q2PPJ7
Gene Alias 250 kDa substrate of Akt; A230067G21Rik; akt substrate AS250; AS250; bA287B20.1; BC053994; C20orf74; dJ1049G11; dJ1049G11.4; Kiaa1272; p220; pp250; Ral GTPase activating protein catalytic alpha subunit 2; Ral GTPase activating protein, alpha subunit 2 (catalytic); ral GTPase-activating protein alpha subunit 2; ral GTPase-activating protein subunit alpha-2; RALGAPA2; RapGAPalpha2; RGC2
Gene Symbols RALGAPA2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RALGAPA2. Recombinant protein control fragment (Product #RP-99469).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57186
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.