Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RALGAPB Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562410
Description
Immunogen sequence: LPCRPFDTVF IFYMKPGQKT NQEILKNVES SRTVQPHFLE FLLSLGWSVD VGRHPGWTGH VSTSWSINCC DDGEGSQQEV ISSEDIGASI FNGQKKVLYY ADAL Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 94%.
Non-catalytic subunit of the heterodimeric RalGAP1 and RalGAP2 complexes which act as GTPase activators for the Ras-like small GTPases RALA and RALB.
Specifications
RALGAPB | |
Polyclonal | |
Unconjugated | |
RALGAPB | |
9330195P16; AI507211; B230339M05Rik; DKFZp781M2411; KIAA1219; mKIAA1219; p170; p170 {ECO:0000303; PubMed:19520869}; Ral GTPase activating protein non-catalytic beta subunit; Ral GTPase activating protein non-catalytic subunit beta; Ral GTPase activating protein, beta subunit (non-catalytic); ral GTPase-activating protein beta subunit; ral GTPase-activating protein subunit beta; ralgapb; ralgapb {ECO:0000303; RalGAPbeta; RGD1306861; zmp:0000000942 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57148 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86X10 | |
RALGAPB | |
Recombinant protein corresponding to Human RALGAPB. Recombinant protein control fragment (Product #RP-99448). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction