Learn More
RALGAPB Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304959A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat
Non-catalytic subunit of the heterodimeric RalGAP1 and RalGAP2 complexes which act as GTPase activators for the Ras-like small GTPases RALA and RALB.Specifications
RALGAPB | |
Polyclonal | |
Unconjugated | |
RALGAPB | |
DKFZp781M2411, KIAA1219, p170, Ral GTPase activating protein, beta subunit (non-catalytic), RalGAPbeta | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
228850, 57148 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q86X10, Q8BQZ4 | |
RALGAPB | |
Between 1025 and 1075 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.