Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RALGDS Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567784
Description
Predicted to react with Mouse and Rat samples.
Stimulates the dissociation of GDP from the Ras-related RalA and RalB GTPases which allows GTP binding and activation of the GTPases. Interacts and acts as an effector molecule for R-Ras, H-Ras, K-Ras, and Rap.
Specifications
RALGDS | |
Polyclonal | |
Unconjugated | |
RALGDS | |
Gnds; KIAA1308; mKIAA1308; Ral guanine nucleotide dissociation stimulator; ral guanine nucleotide exchange factor; RALGDS; RalGEF; RGDS; RGF | |
Rabbit | |
Affinity Chromatography | |
RUO | |
19730, 29622, 5900 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q03385, Q03386, Q12967 | |
RALGDS | |
Synthesized peptide derived from human RALGDS. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction