Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RALY Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297634JF646
Description
RALY Polyclonal specifically detects RALY in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
RALY | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
Autoantigen p542, Heterogeneous nuclear ribonucleoprotein C-like 2, hnRNP associated with lethal yellow protein homolog, hnRNP core protein C-like 2, HNRPCL2MGC117312, P542hnRNP-associated with lethal yellow), RNA binding protein (autoantigenic, hnRNP-associated with lethal yellow), RNA binding protein, autoantigenic (hnRNP-associated with lethal yellow homolog(mouse)), RNA-binding protein (autoantigenic), RNA-binding protein Raly | |
Produced in rabbits immunized with E. coli-derived Human RALY fragment. | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
22913 | |
Store at 4°C in the dark. | |
IgG |
Immunocytochemistry, Immunofluorescence | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction