Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RALY Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530929
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530929 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530929 Supplier Invitrogen™ Supplier No. PA530929
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HepG2. Predicted reactivity: Mouse (93%), Rat (86%), Zebrafish (82%), Dog (86%), Chimpanzee (98%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

In infectious mononucleosis, anti-EBNA-1 antibodies are produced which cross-react with multiple normal human proteins. The cross-reactivity is due to anti-gly/ala antibodies that cross-react with host proteins containing configurations like those in the EBNA-1 repeat. One such antigen is RALY which is a member of the heterogeneous nuclear ribonucleoprotein gene family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RALY
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.64 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene RALY
Gene Accession No. Q64012, Q9UKM9
Gene Alias AI663842; Autoantigen p542; autoantigenic hnRNP-associated with lethal yellow; heterogeneous nuclear ribonucleoprotein C-like 2; hnR; hnRNP associated with lethal yellow protein; hnRNP associated with lethal yellow protein homolog; hnRNP core protein C-like 2; hnRNP-associated with lethal yellow; HNRPCL2; LOW QUALITY PROTEIN: RNA-binding protein Raly; maternally-expressed hnRNP C-related protein; Merc; MGC117312; P542; RALY; RALY heterogeneous nuclear ribonucleoprotein; RALY protein; RNA binding protein; RNA binding protein (autoantigenic, hnRNP-associated with lethal yellow) short isoform; RNA binding protein, autoantigenic (hnRNP-associated with lethal yellow homolog (mouse)); RNA binding protein, autoantigenic (hnRNP-associated with lethal yellow homolog); RNA-binding protein (autoantigenic); RNA-binding protein (autoantigenic, hnRNP-associated with lethal yellow); RNA-binding protein Raly; zgc:103445
Gene Symbols RALY
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RALY. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19383, 22913, 296301
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.