Learn More
Invitrogen™ RALY Polyclonal Antibody

Description
Immunogen sequence: RLFDYRGRLS PVPVPRAVPV KRPRVTVPLV RRVKTNVPVK LFARSTAVTT SSAKIKLKSS ELQAIK Highest antigen sequence indentity to the following orthologs: Rat - 91%, Mouse - 94%.
Specifications
Specifications
| Antigen | RALY |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | RALY |
| Gene Accession No. | Q64012, Q9UKM9 |
| Gene Alias | AI663842; Autoantigen p542; autoantigenic hnRNP-associated with lethal yellow; heterogeneous nuclear ribonucleoprotein C-like 2; hnR; hnRNP associated with lethal yellow protein; hnRNP associated with lethal yellow protein homolog; hnRNP core protein C-like 2; hnRNP-associated with lethal yellow; HNRPCL2; LOW QUALITY PROTEIN: RNA-binding protein Raly; maternally-expressed hnRNP C-related protein; Merc; MGC117312; P542; RALY; RALY heterogeneous nuclear ribonucleoprotein; RALY protein; RNA binding protein; RNA binding protein (autoantigenic, hnRNP-associated with lethal yellow) short isoform; RNA binding protein, autoantigenic (hnRNP-associated with lethal yellow homolog (mouse)); RNA binding protein, autoantigenic (hnRNP-associated with lethal yellow homolog); RNA-binding protein (autoantigenic); RNA-binding protein (autoantigenic, hnRNP-associated with lethal yellow); RNA-binding protein Raly; zgc:103445 |
| Gene Symbols | RALY |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.