Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAM Scientific Safe-T-Fill Blood Gas Capillary Tube Mixing Components
Click to view available options
Product Type:
Tube Mixing Fleas
Tube Mixing Magnet
Description
Magnetic rings and mixing fleas (mixing components) for SAFE-T-FILL™ Plastic Capillary Blood Gas Tubes prevent clotting in blood gas capillary tube samples.
- Magnetic Rings measure Ø 8.8mm (width) x 3.9mm (length) ; hole diameter Ø 3.9mm
- Mixing Fleas measure Ø 6.7mm (width) x 12mm (length)
- Packaging labels are marked with Lot Number
Specifications
Specifications
| Product Type | Tube Mixing Magnet |
| Material | Ferrite Magnet (Ceramic) |
| Sterility | Non-sterile |
| For Use With (Equipment) | Capillary Blood Collection Tubes |
| For Use With (Application) | Intended to incorporate the blood sample with the anticoagulant by moving the mixing wire from side to side with the magnetic ring. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction