Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RAMP2 Antibody (OTI4E5), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273796H
Description
RAMP2 Monoclonal specifically detects RAMP2 in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry.Specifications
RAMP2 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
RAMP2 | |
Full length recombinant protein of human RAMP2 (NP_005845) produced in HEK293T cells. | |
0.1 mL | |
10266.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI4E5 | |
Western Blot, Immunohistochemistry | |
calcitonin receptor-like receptor activity modifying protein 2, Calcitonin-receptor-like receptor activity-modifying protein 2, CRLR activity-modifying protein 2, receptor (calcitonin) activity modifying protein 2, receptor (G protein-coupled) activity modifying protein 2, receptor activity modifying protein 2, receptor activity-modifying protein 2, receptor-activity-modifying protein 2 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Monkey | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction