Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAMP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110266
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110266 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110266 Supplier Invitrogen™ Supplier No. PA5110266
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MASLRVERAG GPRLPRTRVG RPAALRLLLL LGAVLNPHEA LAQPLPTTGT PGSEGGTVKN YETAVQFCWN HYKDQMDPIE KDWCDWAMIS RPYSTLRDCL EHFAELFDLG FPNPLAERII FETHQIHFAN CSLVQPTFSD PPEDV.

The protein encoded by this gene is a member of the RAMP family of single-transmembrane-domain proteins, called receptor (calcitonin) activity modifying proteins (RAMPs). RAMPs are type I transmembrane proteins with an extracellular N terminus and a cytoplasmic C terminus. RAMPs are required to transport calcitonin-receptor-like receptor (CRLR) to the plasma membrane. CRLR, a receptor with seven transmembrane domains, can function as either a calcitonin-gene-related peptide (CGRP) receptor or an adrenomedullin receptor, depending on which members of the RAMP family are expressed. In the presence of this (RAMP2) protein, CRLR functions as an adrenomedullin receptor. The RAMP2 protein is involved in core glycosylation and transportation of adrenomedullin receptor to the cell surface.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAMP2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.73 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Ramp2
Gene Accession No. O60895, Q9JHJ1, Q9WUP0
Gene Alias calcitonin receptor-like receptor activity modifying protein 2; calcitonin-receptor-like receptor activity-modifying protein 2; CRLR activity-modifying protein 2; OTTMUSP00000002615; OTTMUSP00000037624; Ramp2; receptor (calcitonin) activity modifying protein 2; receptor (G protein-coupled) activity modifying protein 2; receptor activity modifying protein 2; receptor activity modifying protein 2 isoform; receptor activity-modifying protein 2; receptor-activity-modifying protein 2; RP23-281C18.6
Gene Symbols Ramp2
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10266, 54409, 58966
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.