Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAMP3 Antibody [Alexa Fluor™Plus 680] - Humanized, R∓D Systems™
SDP

Catalog No. RD10083223 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RD10083223 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RD10083223 Supplier R&D Systems Supplier No. NBP327994AFP680

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Human Monoclonal Antibody

RAMP3 Monoclonal antibody specifically detects RAMP3 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay

Specifications

Antigen RAMP3
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Conjugate Alexa Fluor Plus 680
Formulation 50mM Sodium Borate
Gene Alias calcitonin receptor-like receptor activity modifying protein 3, Calcitonin-receptor-like receptor activity-modifying protein 3, CRLR activity-modifying protein 3, receptor (calcitonin) activity modifying protein 3, receptor (G protein-coupled) activity modifying protein 3, receptor activity modifying protein 3, receptor activity-modifying protein 3, receptor-activity-modifying protein 3
Host Species Human
Immunogen RAMP3
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10268
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.