Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAMP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5145110
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5145110 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5145110 Supplier Invitrogen™ Supplier No. PA5145110
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SFTNCTEMEA NVVGCYWPNP LAQGFITGIH RQFFSNCTVD RVHLEDPPDE V Antigen sequence identity: Mouse 88%; Rat 90%.

The protein encoded by this gene is a member of the RAMP family of single-transmembrane-domain proteins, called receptor (calcitonin) activity modifying proteins (RAMPs). RAMPs are type I transmembrane proteins with an extracellular N terminus and a cytoplasmic C terminus. RAMPs are required to transport calcitonin-receptor-like receptor (CRLR) to the plasma membrane. CRLR, a receptor with seven transmembrane domains, can function as either a calcitonin-gene-related peptide (CGRP) receptor or an adrenomedullin receptor, depending on which members of the RAMP family are expressed. In the presence of this (RAMP3) protein, CRLR functions as an adrenomedullin receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAMP3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Ramp3
Gene Accession No. O60896
Gene Alias AI850306; calcitonin receptor-like receptor activity modifying protein 3; calcitonin-receptor-like receptor activity-modifying protein 3; CRLR activity-modifying protein 3; Ramp3; receptor (calcitonin) activity modifying protein 3; receptor (G protein-coupled) activity modifying protein 3; receptor activity modifying protein 3; receptor activity-modifying protein 3; receptor-activity-modifying protein 3
Gene Symbols Ramp3
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 68-118. Recombinant protein control fragment (Product #RP-110144).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10268
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.