Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RAN Mouse anti-Human, Monkey, Rat, Clone:8D1H12, Abnova™

Catalog No. 89333591 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-591 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-591 Supplier Abnova Corporation Supplier No. MAB17210
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human RAN.

RAN (ras-related nuclear protein) is a small GTP binding protein belonging to the RAS superfamily that is essential for the translocation of RNA and proteins through the nuclear pore complex. The RAN protein is also involved in control of DNA synthesis and cell cycle progression. Nuclear localization of RAN requires the presence of regulator of chromosome condensation 1 (RCC1). Mutations in RAN disrupt DNA synthesis. Because of its many functions, it is likely that RAN interacts with several other proteins. RAN regulates formation and organization of the microtubule network independently of its role in the nucleus-cytosol exchange of macromolecules. RAN could be a key signaling molecule regulating microtubule polymerization during mitosis. RCC1 generates a high local concentration of RAN-GTP around chromatin which, in turn, induces the local nucleation of microtubules. RAN is an androgen receptor (AR) coactivator that binds differentially with different lengths of polyglutamine within the androgen receptor. Polyglutamine repeat expansion in the AR is linked to Kennedy′s disease (X-linked spinal and bulbar muscular atrophy). RAN coactivation of the AR diminishes with polyglutamine expansion within the AR, and this weak coactivation may lead to partial androgen insensitivity during the development of Kennedy′s disease. [provided by RefSeq]

Specifications

Antigen RAN
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 8D1H12
Conjugate Unconjugated
Description RAN, member RAS oncogene family
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunohistochemistry (1:200-1:1000)
Immunocytochemistry
Flow Cytometry (1:200-1:400)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene RAN
Gene Alias ARA24, Gsp1, TC4
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 1-216 of human RAN from E. coli.
Quantity 100 μg
Gene ID (Entrez) 5901
Target Species Human, Monkey, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.