Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RAN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598111
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598111 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598111 Supplier Invitrogen™ Supplier No. PA598111
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RAN Polyclonal Antibody for Western Blot, IHC (P), ELISA

Ran (ras-related nuclear protein) is a small GTP binding protein belonging to the RAS superfamily that is essential for the translocation of RNA and proteins through the nuclear pore complex. The Ran protein is also involved in control of DNA synthesis and cell cycle progression. Nuclear localization of Ran requires the presence of regulator of chromosome condensation 1 (RCC1).
TRUSTED_SUSTAINABILITY

Specifications

Antigen RAN
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RAN
Gene Accession No. P62826
Gene Alias AAF30287; androgen receptor-associated protein 24; ARA24; ARA25; CG1404; CG1404-PA; CG1404-PB; CG1404-PC; CG1404-PD; Chromosome stability protein 17; CNR1; CST17; Dmel\CG1404; Dmel_CG1404; dran; Genetic suppressor of PRP20-1; GM14354p; gsp1; GTPase Ran; GTPase Ran homolog; GTP-binding nuclear protein GSP1/CNR1; GTP-binding nuclear protein Ran; guanosine triphosphatase Ran; I79_024397; l(1)G0075; L8003.19; LD32416p; LD40852p; member RAS oncogene family; OK/SW-cl.81; OTTHUMP00000166078; OTTHUMP00000166102; Q9VZ23; ran; Ran GTPase; Ran GTPase GSP1; ran GTP-binding protein; RAN, member RAS oncogene family; RAN, member RAS oncogene family L homeolog; ran.L; ran-1; Ran10A; RanGTP; RanGTPase; Ran-PA; Ran-PB; Ran-PC; Ran-PD; RanTC4; Rasl2-8; Ras-like protein TC4; ras-related nuclear protein; tc4; XELAEV_18007741mg; xran; YLR293C
Gene Symbols RAN
Host Species Rabbit
Immunogen Recombinant Human GTP-binding nuclear protein Ran protein (1-216AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5901
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.