Learn More
Ran Rabbit anti-Human, Polyclonal, Bioss

Description
Specifications
Specifications
| Antigen | Ran |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH |
| Gene | RAN |
| Gene Accession No. | P62826 |
| Gene Alias | AAF30287; Androgen receptor-associated protein 24; ARA24; ARA25; CG1404; CG1404 gene product from transcript CG1404-RC; CG1404-PA; CG1404-PB; CG1404-PC; CG1404-PD; Dmel\CG1404; Dmel_CG1404; dran; Gsp1; GTPase Ran; GTP-binding nuclear protein Ran; guanosine triphosphatase Ran; I79_024397; l(1)G0075; member RAS oncogene family; OK/SW-cl.81; OTTHUMP00000166078; OTTHUMP00000166102; Q9VZ23; ran; Ran GTPase; ran GTP-binding protein; RAN, member RAS oncogene family; RAN, member RAS oncogene family L homeolog; ran.L; ran-1; Ran10A; RanGTP; RanGTPase; Ran-PA; Ran-PB; Ran-PC; Ran-PD; RanTC4; Rasl2-8; Ras-like protein TC4; Ras-related nuclear protein; TC4; XELAEV_18007741mg; xran |
| Gene Symbols | RAN |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.