Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RanBP1 Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10079815FR
Description
RanBP1 Polyclonal antibody specifically detects RanBP1 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
RanBP1 | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
HTF9A, MGC88701, RAN binding protein 1, Ran-binding protein 1, RanBP1, ran-specific GTPase-activating protein | |
The immunogen recognized by this antibody maps to a region between residue 150 and the C-terminus (residue 201) of human RAN Binding Protein 1 using the numbering given in entry NP_002873.1 (GeneID 5902). | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
5902 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction