Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RanBP1 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001443 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001443 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001443 Supplier Novus Biologicals Supplier No. NB1001443
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

RanBP1 Polyclonal specifically detects RanBP1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen RanBP1
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.03 - 0.5 ug/mL, Flow Cytometry 10 ug/mL, Immunohistochemistry 3 - 5 ug/mL, Immunocytochemistry/Immunofluorescence 10 ug/mL, Immunohistochemistry-Paraffin 3 - 5 ug/mL, Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_002873.
Gene Alias HTF9A, MGC88701, RAN binding protein 1, Ran-binding protein 1, RanBP1, ran-specific GTPase-activating protein
Gene Symbols RANBP1
Host Species Goat
Immunogen Peptide with sequence C-LSVKEETKEDAEEKQ corresponding to C-Terminus according to NP_002873.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 5902
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.