Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RanBP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | RanBP1 |
---|---|
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
RanBP1 Polyclonal specifically detects RanBP1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
RanBP1 | |
Polyclonal | |
Rabbit | |
DNA replication Transcription Translation and Splicing | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
5902 | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:AKDTHEDHDTSTENTDESNHDPQFEPIVSL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human | |
HTF9A, MGC88701, RAN binding protein 1, Ran-binding protein 1, RanBP1, ran-specific GTPase-activating protein | |
RANBP1 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title