Learn More
RANBP1 Mouse anti-Human, Clone: OTI8E1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF811554

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Ran/TC4-binding protein, RanBP1, interacts specifically with GTP-charged RAN. RANBP1 encodes a 23-kD protein that binds to RAN complexed with GTP but not GDP. RANBP1 does not activate GTPase activity of RAN but does markedly increase GTP hydrolysis by the RanGTPase-activating protein (RanGAP1). The RANBP1 cDNA encodes a 201-amino acid protein that is 92% similar to its mouse homolog. In both mammalian cells and in yeast, RANBP1 acts as a negative regulator of RCC1 by inhibiting RCC1-stimulated guanine nucleotide release from RAN.Specifications
RANBP1 | |
Monoclonal | |
Unconjugated | |
RANBP1 | |
HTF9A | |
Mouse | |
Affinity Chromatography | |
RUO | |
5902 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI8E1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P43487 | |
RANBP1 | |
Full length human recombit protein of human RANBP1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.