Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RANBP10 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177372MFV610
Description
RANBP10 Polyclonal antibody specifically detects RANBP10 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
RANBP10 | |
Polyclonal | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
KIAA1464FLJ31165, RAN binding protein 10, ran-binding protein 10, RanBP10 | |
Antibody was raised against a 15 amino acid synthetic peptide near the center of human RANBP10. The immunogen is located within amino acids 380 - 430 of RANBP10. Amino Acid Squence: VASTSKQNHSKYPA | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
57610 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction