Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RANBP10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520930
Description
A suggested positive control is human skeletal muscle tissue lysate. PA5-20930 can be used with blocking peptide PEP-1044.
The RAN binding protein 10 (RANBP10) is a cytoplasmic guanine nucleotide exchange factor that shares high sequence similarity to the MET-interacting protein RANBP9. RANBP10 is expressed in the cytoplasm of mature megakaryocytes (MKs) and platelets where it localizes to polymerized noncentrosomal microtubules. RANBP10 possesses guanine nucleotide exchange factor activity towards RAN, a GTPase involved in nucleocytoplasmic traffic, spindle formation at mitosis, and nuclear envelope assembly during telophase, suggesting that RANBP10 may also be involved in these activities. RANBP10 depletion in vitro caused the disturbance of polymerized filaments in MKs and platelets in RANBP10-null mice exhibited disorders in microtubule filament numbers and localization, indicating that RANBP10 may play a significant role in maintaining microtubule dynamics.
Specifications
| RANBP10 | |
| Polyclonal | |
| Unconjugated | |
| RANBP10 | |
| 4432417N03Rik; KIAA1464; RAN binding protein 10; ran-binding protein 10; RANBP10; rangef; RGD1311587 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 57610, 74334 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q6VN19, Q6VN20 | |
| RANBP10 | |
| A 15 amino acid peptide near the center of human RANBP10. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction