Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RANBP10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110267
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110267 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110267 Supplier Invitrogen™ Supplier No. PA5110267
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YPGSPSLSPR HGPSSSHMHN TGADSPSCSN GVASTKSKQN HSKYPAPSSS SSSSSSSSSS SPSSVNYSES NSTDSTKSQH HSSTSNQETS DSEMEMEAEH YPNGVLGSMS TRIVNGAYKH EDLQTDESSM DDRHPR.

The RAN binding protein 10 (RANBP10) is a cytoplasmic guanine nucleotide exchange factor that shares high sequence similarity to the MET-interacting protein RANBP9. RANBP10 is expressed in the cytoplasm of mature megakaryocytes (MKs) and platelets where it localizes to polymerized noncentrosomal microtubules. RANBP10 possesses guanine nucleotide exchange factor activity towards RAN, a GTPase involved in nucleocytoplasmic traffic, spindle formation at mitosis, and nuclear envelope assembly during telophase, suggesting that RANBP10 may also be involved in these activities. RANBP10 depletion in vitro caused the disturbance of polymerized filaments in MKs and platelets in RANBP10-null mice exhibited disorders in microtubule filament numbers and localization, indicating that RANBP10 may play a significant role in maintaining microtubule dynamics.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RANBP10
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.59 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene RANBP10
Gene Accession No. Q6VN19, Q6VN20
Gene Alias 4432417N03Rik; KIAA1464; RAN binding protein 10; ran-binding protein 10; RANBP10; rangef; RGD1311587
Gene Symbols RANBP10
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 362-497 of human RANBP10 (NP_065901.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57610, 74334
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.