Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RanBP16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111268
Change view
Click to view available options
Quantity:
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111268 100 μL
PIPA5111187 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA5111268 Supplier Invitrogen™ Supplier No. PA5111268
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SQPPEKQQAM HLCFENLMEG IERNLLTK Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.

The transport of protein and large RNAs through the nuclear pore complexes is an energy-dependent and regulated process. The import of proteins with a nuclear localization signal is accomplished by recognition of one or more clusters of basic amino acids by the importin-alpha/beta complex; see MIM 600685 and MIM 602738. The small GTPase RAN plays a key role in NLS-dependent protein import. RAN-binding protein-16 is a member of the importin-beta superfamily of nuclear transport receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RanBP16
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Xpo7
Gene Accession No. Q9UIA9
Gene Alias 4930506C02Rik; BB164534; EXP7; exportin 7; exportin-7; Kiaa0745; mKIAA0745; RAN binding protein 16; ran-binding protein 16; Ranbp16; Xpo7
Gene Symbols Xpo7
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human RanBP16. Recombinant protein control fragment (Product #RP-100394).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23039
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.