Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RANBP17 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$254.42 - $665.78
Specifications
| Antigen | RANBP17 |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml |
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
RANBP17 Polyclonal specifically detects RANBP17 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| RANBP17 | |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 64901 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:MDGELSCRVFQLISLMDTGLPRCCNEKIELAILWFLDQFRKTYVGDQLQRTSKVYARMSEVLGITDDNHVLETFMT | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Human | |
| FLJ32916, RAN binding protein 17 | |
| RANBP17 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title