Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RANBP17 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556193
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556193 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556193 Supplier Invitrogen™ Supplier No. PA556193
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDGELSCRVF QLISLMDTGL PRCCNEKIEL AILWFLDQFR KTYVGDQLQR TSKVYARMSE VLGITDDNHV LETFMT Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

The transport of protein and large RNAs through the nuclear pore complexes (NPC) is an energy-dependent and regulated process. The import of proteins with a nuclear localization signal (NLS) is accomplished by recognition of one or more clusters of basic amino acids by the importin-alpha/beta complex; see MIM 600685 and MIM 602738. The small GTPase RAN (MIM 601179) plays a key role in NLS-dependent protein import. RAN-binding protein-17 is a member of the importin-beta superfamily of nuclear transport receptors.[supplied by OMIM, Jul 2002].
TRUSTED_SUSTAINABILITY

Specifications

Antigen RANBP17
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RANBP17
Gene Accession No. Q9H2T7
Gene Alias RAN binding protein 17; ran-binding protein 17; Ranbp17
Gene Symbols RANBP17
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RANBP17. Recombinant protein control fragment (Product #RP-91863).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64901
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.