Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RanBP2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB1202938
Description
RanBP2 Polyclonal specifically detects RanBP2 in Human, Mouse, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RanBP2 | |
Polyclonal | |
Unconjugated | |
PBS containing 1 mg/ml BSA with 0.05% Sodium Azide | |
ANE1, E3 SUMO-protein ligase RanBP2, EC 5.2.1.8, Nuclear pore complex protein Nup358, nucleoporin 358, Nucleoporin Nup358, NUP358TRP2, P270, RAN binding protein 2, Ran-binding protein 2, RanBP2, transformation-related protein 2,358 kDa nucleoporin, TRP1 | |
Rabbit | |
Affinity Purified | |
RUO | |
5903 | |
Human, Bovine | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Western Blot 1 ug/ml, Immunohistochemistry 10 ug/ml, Immunohistochemistry-Paraffin 1:10 - 1:500 | |
P49792 | |
RANBP2 | |
Synthetic peptide corresponding to residues M(1) R R S K A D V E R Y I A S V Q G S(18) of human RanBP2. | |
100 μg | |
Primary | |
Human and Cow. Not yet tested in other species. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction