Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RanBP3 Antibody (12E11), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242672R
Description
RanBP3 Monoclonal specifically detects RanBP3 in Human, Mouse samples. It is validated for Western Blot, ELISA.Specifications
RanBP3 | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RANBP3 | |
Recombinant human RANBP3 (235-445aa) purified from E. coli | |
100 μL | |
Signal Transduction | |
8498 | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, ELISA | |
1200000000000 | |
Western Blot, ELISA | |
DKFZp586I1520, RAN binding protein 3, ran-binding protein 3, RAN-binding protein-3, RanBP3 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction