Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RANBP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1084
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1084 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1084 Supplier Invitrogen™ Supplier No. PA1084
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-084 detects Ran binding protein 3 (RanBP3) from human and rat cells. PA1-084 has been successfully used in Western blot and immunofluorescence procedures. By Western blot this antibody detects an ~60 kDa protein representing RanBP3 from HeLa cell extracts. Immunofluorescence staining of RanBP3 in NRK cells with PA1-084 results in diffuse nuclear staining. The PA1-084 immunogen is a synthetic peptide corresponding to residues M(1) A D L A N E E K P A L A(13) C of human RanBP3. The PA1-084 immunizing peptide (Cat. # PEP-177) is available for use in neutralization and control experiments.

Ran binding protein 3 (RanBP3) is a member of Ran GTPase binding proteins that is located at the nuclear pore complex (NPC) and plays an essential role in the transport between cytoplasm and nucleus. RANPB3 is expessed as two spliced forms, RanBP3a and RanBP3b, both of which contains two FXFG motifs, characteristic of a subgroup of nucleoporins in addition to nuclear localization signal in the amino-terminal region (amino acid residues 40-57) and a unique acidic carboxyl terminus which is integral to its switch function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RANBP3
Applications Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1 mg/mL BSA and 0.05% sodium azide
Gene RANBP3
Gene Accession No. Q9H6Z4
Gene Alias 2610024N24Rik; AA408221; RAN binding protein 3; RANB3; ran-binding protein 3; RAN-binding protein-3; Ranbp3
Gene Symbols RANBP3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues M(1) A D L A N E E K P A L A(13) C of human RanBP3.
Purification Method Antigen affinity chromatography
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 501281, 8498
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.