Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RANBP3 Recombinant Rabbit Monoclonal Antibody (8H7W3)
GREENER_CHOICE

Catalog No. PIMA542584
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542584 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542584 Supplier Invitrogen™ Supplier No. MA542584
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

The target is usually found in the following locations: Cytoplasm, Nucleus. Immunogen sequence: MADLANEEKP AIAPPVFVFQ KDKGQKSPAE QKNLSDSGEE PRGEAEAPHH GTGHPESAGE HALEPPAPAG ASASTPPPPA PEAQLPPFPR ELAGRSAGGS.

Ran binding protein 3 (RanBP3) is a member of Ran GTPase binding proteins that is located at the nuclear pore complex (NPC) and plays an essential role in the transport between cytoplasm and nucleus. RANPB3 is expessed as two spliced forms, RanBP3a and RanBP3b, both of which contains two FXFG motifs, characteristic of a subgroup of nucleoporins in addition to nuclear localization signal in the amino-terminal region (amino acid residues 40-57) and a unique acidic carboxyl terminus which is integral to its switch function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RANBP3
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone 8H7W3
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RANBP3
Gene Accession No. Q9CT10
Gene Alias 2610024N24Rik; AA408221; RAN binding protein 3; RANB3; ran-binding protein 3; RAN-binding protein-3; Ranbp3
Gene Symbols RANBP3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human RANBP3 (Q9H6Z4).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 501281, 71810
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.