Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RANBP6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA537033
Description
This antibody detects endogenous protein at a molecular weight of 125 kDa. Purity is >95% by SDS-PAGE.
RANBP6 (Ran-binding protein 6) belongs to the importin beta family and may function in nuclear protein import as a nuclear transport receptor.
Specifications
RANBP6 | |
Polyclonal | |
Unconjugated | |
RANBP6 | |
C630001B19; RAN binding protein 6; Ran BP6; ran-binding protein 6; Ranbp6; Ran-BP6; Ran-GTP binding protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
240614, 26953, 309326 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O60518, Q8BIV3 | |
RANBP6 | |
Synthetic peptide corresponding to amino acids 841-886 of human RANBP6. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction