Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RANBP6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584120
Description
Immunogen sequence: LSSGFEEVYP NLPADVQRDV KIELILAVKL ETHA.
RANBP6 (Ran-binding protein 6) belongs to the importin beta family and may function in nuclear protein import as a nuclear transport receptor.
Specifications
RANBP6 | |
Polyclonal | |
Unconjugated | |
RANBP6 | |
C630001B19; RAN binding protein 6; Ran BP6; ran-binding protein 6; Ranbp6; Ran-BP6; Ran-GTP binding protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26953 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60518 | |
RANBP6 | |
Recombinant protein corresponding to Human RANBP6. Recombinant protein control fragment (Product #RP-106408). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction