Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RANBP9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139949
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139949 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139949 Supplier Invitrogen™ Supplier No. PA5139949
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: ELNSINMSRS QQVNNFTSND VDMETDHYSN GVGETSSNGF LNGSSKHDHE MEDCDTVMEV DSS Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 95%.

This gene encodes a protein that binds RAN, a small GTP binding protein belonging to the RAS superfamily that is essential for the translocation of RNA and proteins through the nuclear pore complex. The protein encoded by this gene has also been shown to interact with several other proteins, including met proto-oncogene, homeodomain interacting protein kinase 2, androgen receptor, and cyclin-dependent kinase 11.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RANBP9
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RANBP9
Gene Accession No. Q96S59
Gene Alias B cell antigen receptor Ig beta associated protein 1; B-cell antigen receptor Ig beta-associated protein 1; BPM 90; BPM L; BPM90; BPML; BPM-L; IBAP 1; Ibap1; IBAP-1; Imp 9; Importin 9; novel centrosomal protein RanBPM; RAN binding protein 9; Ran Binding Protein in the Microtubule organizing center; ran binding protein, centrosomal; Ran BP9; ran-binding protein 9; Ran-binding protein M; RanBP 7; RANBP 9; RanBP7; Ranbp9; RANBPM; RGD1561940
Gene Symbols RANBP9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RANBP9. Recombinant protein control fragment (Product #RP-109590).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10048
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.