Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ EvenCoat Streptavidin Coated Plates
GSA_VA

Catalog No. p-4758024
Click to view available options
Quantity:
15/Pk.
5/Pk.
This item is not returnable. View return policy

Catalog No. CP004

Supplier: R&D Systems™ CP004

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

used as a solid support for assays that utilize a biotinylated molecule or otherwise require the binding of biotin.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Color Clear
Concentration 4 μg/mL
Quantity 15/Pk.
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
For Use With (Application) ELISA
Specificity Biotinylated molecules
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.