Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RanGAP1 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP118915V
Description
RanGAP1 Polyclonal antibody specifically detects RanGAP1 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
RanGAP1 | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
KIAA1835Fug1, MGC20266, Ran GTPase activating protein 1, ran GTPase-activating protein 1, RanGAP1, SD, segregation distorter homolog, segregation distorter homolog (Drosophila), segregation distortion | |
The immunogen recognized by this antibody maps to a region between residue 100 and 150 of human Ran GTPase activating protein 1 using the numbering given in entry NP_002874.1 (GeneID 5905). | |
0.1 mL | |
Cellular Markers | |
5905 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction