Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RANGAP1 Mouse anti-Human, Clone:4F9E2, Abnova™

Catalog No. 89333188 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-188 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-188 Supplier Abnova Corporation Supplier No. MAB16611
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human RANGAP1.

RanGAP1, is a homodimeric 65-kD polypeptide that specifically induces the GTPase activity of RAN, but not of RAS by over 1,000-fold. RanGAP1 is the immediate antagonist of RCC1, a regulator molecule that keeps RAN in the active, GTP-bound state. The RANGAP1 gene encodes a 587-amino acid polypeptide. The sequence is unrelated to that of GTPase activators for other RAS-related proteins, but is 88% identical to Fug1, the murine homolog of yeast Rna1p. RanGAP1 and RCC1 control RAN-dependent transport between the nucleus and cytoplasm. RanGAP1 is a key regulator of the RAN GTP/GDP cycle. [provided by RefSeq]

Specifications

Antigen RANGAP1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4F9E2
Conjugate Unconjugated
Description Ran GTPase activating protein 1
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene RANGAP1
Gene Alias Fug1, KIAA1835, MGC20266, SD
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 359-587 of human RANGAP1 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 5905
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.