Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RANGRF Monoclonal Antibody (1H4)
GREENER_CHOICE

Catalog No. PIMA524454
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524454 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524454 Supplier Invitrogen™ Supplier No. MA524454
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MEPTRDCPLF GGAFSAILPM GAIDVSDLRP VPDNQEVFCH PVTDQSLIVE LLELQAHVRG EAAARYHFED VGGVQGARAV HVESVQPLSL ENLALRGRCQ EAWVLSGKQQ IAKENQQVAK DVTLHQALLR LPQYQTDLLL TFNQPP.

May regulate the intracellular trafficking of RAN. In cardiac cells seems to regulate the cell surface localization of SCN5A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RANGRF
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1H4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene RANGRF
Gene Accession No. Q9HD47
Gene Alias 2400006H24Rik; HSPC165; HSPC236; MDS5; MOG1; MOG1 homolog; ran guanine nucleotide release factor; Ran-binding protein MOG1; Rangnrf; RANGRF
Gene Symbols RANGRF
Host Species Mouse
Immunogen RANGRF (AAI00018.1, 1 a.a. ~ 146 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29098
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.