Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RANGRF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563501
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563501 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563501 Supplier Invitrogen™ Supplier No. PA563501
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VAKDVTLHQA LLRLPQYQTD LLLTFNQPPP DNRSSLGPEN LSPAPWSLGD FEQLVTSLTL HDPNIFGP Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.

May regulate the intracellular trafficking of RAN. In cardiac cells seems to regulate the cell surface localization of SCN5A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RANGRF
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RANGRF
Gene Accession No. Q9HD47
Gene Alias 2400006H24Rik; HSPC165; HSPC236; MDS5; MOG1; MOG1 homolog; ran guanine nucleotide release factor; Ran-binding protein MOG1; Rangnrf; RANGRF
Gene Symbols RANGRF
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RANGRF. Recombinant protein control fragment (Product #RP-93218).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29098
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.